Learn More
Aviva Systems Biology Corp Nmral1 Antibody - N-terminal region (ARP37361_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37361_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nmral1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the n terminal region of mouse Nmral1. Alternate names: 1110025F24Rik, AI256624ApplicationWB. Species reactivity: Cow, Dog, Mouse, Pig, Rat
Catalog No. 50-284-619