Learn More
Aviva Systems Biology Corp NMRAL1 Antibody - middle region (ARP58502_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58502_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NMRAL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NMRAL1. Alternate names: FLJ25918, HSCARG, SDR48A1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62329