Learn More
Medchemexpress LLC HY-15828 5mg Medchemexpress, NMS-1286937 CAS:1034616-18-6 Purity:>98%
SDP
Catalog No. 502030687
Encompass_Preferred
Medchemexpress, HY-15828 5mg NMS-1286937 CAS:1034616-18-6 NMS-1286937 is a potent, selective and orally available PLK1 inhibitor, with an IC50 of 2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0687 Supplier Medchemexpress LLC Supplier No. HY158285MG
Medchemexpress, HY-15828 5mg NMS-1286937 CAS:1034616-18-6 NMS-1286937 is a potent, selective and orally available PLK1 inhibitor, with an IC50 of 2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.