Learn More
Medchemexpress LLC HY-15714 5mg Medchemexpress, NMS-859 CAS:1449236-96-7 Purity:>98%
SDP
Catalog No. 502081775
Encompass_Preferred
Medchemexpress, HY-15714 5mg NMS-859 CAS:1449236-96-7 NMS-859 is a potent, covalent VCP (p97) inhibitor, with IC50s of 0.37 and 0.36 μM for wild-type VCP in the presence of 60 μM and 1 mM ATP in cells, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1775 Supplier Medchemexpress LLC Supplier No. HY157145MG
Medchemexpress, HY-15714 5mg NMS-859 CAS:1449236-96-7 NMS-859 is a potent, covalent VCP (p97) inhibitor, with IC50s of 0.37 and 0.36 μM for wild-type VCP in the presence of 60 μM and 1 mM ATP in cells, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.