Learn More
Selleck Chemical LLC NMS-873
SDP
Supplier:  Selleck Chemical LLC S72851G
Encompass_Preferred
NMS-873 is an allosteric and specific p97 inhibitor with IC50 of 30 nM that demonstrates potent selectivity for VCP/p97 compared to a panel of other AAA ATPases Hsp90 and 53 additional analyzed kinases (IC50s 10 M)
Catalog No. 50-000-04201