Learn More
Selleck Chemical LLC NMS-873 5mg 1418013-75-8
SDP
Supplier:  Selleck Chemical LLC S72855MG
Encompass_Preferred
NMS-873 is an allosteric and specific p97 inhibitor with IC50 of 30 nM that demonstrates potent selectivity for VCP/p97 compared to a panel of other AAA ATPases, Hsp90, and 53 additional analyzed kinases (IC50s >10 ?M). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1147220