Learn More
Selleck Chemical LLC NMS-E973
SDP
Catalog No. 5000002932
Encompass_Preferred
NMS-E973 is a potent and selective Hsp90 inhibitor with DC50 of 10 nM for Hsp90 binding no activiy against a panel of 52 diverse protein kinases
Catalog No. 50-000-02932 Supplier Selleck Chemical LLC Supplier No. S72825MG
NMS-E973 is a potent and selective Hsp90 inhibitor with DC50 of 10 nM for Hsp90 binding no activiy against a panel of 52 diverse protein kinases