Learn More
TARGETMOL CHEMICALS INC NMS-E973 5MG
SDP
Catalog No. 502579326
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NMS-E973 is a potent and selective Hsp90 inhibitor with DC50 of <10 nM for Hsp90 binding no activiy against a panel of 52 diverse protein kinases. purity: 99%
Catalog No. 50-257-9326 Supplier TARGETMOL CHEMICALS INC Supplier No. T66095MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NMS-E973 is a potent and selective Hsp90 inhibitor with DC50 of <10 nM for Hsp90 binding no activiy against a panel of 52 diverse protein kinases. purity: 99%