Learn More
Medchemexpress LLC HY-12382 5mg Medchemexpress, NMS-P715 CAS:1202055-32-0 Purity:>98%
SDP
Catalog No. 502081027
Encompass_Preferred
Medchemexpress, HY-12382 5mg NMS-P715 CAS:1202055-32-0 NMS-P715 is a selective, ATP-competitive inhibitor of MPS1, with an IC50 of 182 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1027 Supplier Medchemexpress LLC Supplier No. HY123825MG
Medchemexpress, HY-12382 5mg NMS-P715 CAS:1202055-32-0 NMS-P715 is a selective, ATP-competitive inhibitor of MPS1, with an IC50 of 182 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.