Learn More
Selleck Chemical LLC Onvansertib (NMS-P937) 10mg 1034616-18-6 PCM-075, NMS1286937
SDP
Supplier:  Selleck Chemical LLC S725510MG
Encompass_Preferred
Onvansertib (NMS-P937, PCM-075, NMS1286937) is an orally available, selective Polo-like Kinase 1 (PLK1) inhibitor with IC50 of 2 nM, 5000-fold selectivity over PLK2/PLK3. Onvansertib (NMS-P937) potently causes a mitotic cell-cycle arrest followed by apoptosis in cancer cell lines and inhibits tumor growth. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4558