Learn More
Proteintech Group Inc NMT1 Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 115461AP150UL
Encompass_Preferred
The NMT1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NMT1. This antibody recognizes human, mouse, rat antigen. The NMT1 antibody has been validated for the following applications: IF, IHC, IP, WB,ELISA analysis.
Catalog No. 50-225-6372