Learn More
Aviva Systems Biology Corp NMUR1 Antibody - C-terminal region (ARP65093_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65093_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NMUR1. It was validated on Western Blot. . Alternate names: (FM-3), FM-3, FM3, GPC-R, GPR66, NMU1RApplicationWB. Species reactivity: Human
Catalog No. 50-288-428