Learn More
Aviva Systems Biology Corp NMUR2 Antibody - N-terminal region (ARP35300_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35300_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NMUR2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NMUR2. Alternate names: FM4, FM-4, TGR1, NMU2R, TGR-1, NMU-R2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55692