Learn More
Sigma Aldrich Fine Chemicals Biosciences NN-Dimethylacetamide Reage
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 1858842.5L
Encompass
N N-Dimethylacetamide (DMAc) is widely used to compose the solvent mixtures for cellulose. Static and dynamic light scattering studies of the cellulose dissolved in varying concentrations of DMAc/LiCl mixtures have been reported.
Catalog No. 50-289-8085