Learn More
Sigma Aldrich Fine Chemicals Biosciences NN-Dimethylformamide Reage1L
Catalog No. 502898137
Encompass
N N-Dimethylformamide (DMf) is a polar organic solvent with a wide range of industrial applications. It is a precursor for numerous reactions such as formylation aminocarbonylation amination amidation and cyanation. Along with phosphorus oxychloride it forms Vilsmeier reagent used in the formylation of reactive aromatic and heteroaromatic substrates. Dielectric relaxation experiments on DMf have been conducted between 238.15 K and 338.15K. The thermophysical properties of the molecular interactions between DMf and the ionic liquids ammonium salts and imidazolium salts have been studied. The characteristics of solute-solvent interactions between ammonium nitrate and DMf have been investigated.
Catalog No. 50-289-8137 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D1585501L
N N-Dimethylformamide (DMf) is a polar organic solvent with a wide range of industrial applications. It is a precursor for numerous reactions such as formylation aminocarbonylation amination amidation and cyanation. Along with phosphorus oxychloride it forms Vilsmeier reagent used in the formylation of reactive aromatic and heteroaromatic substrates. Dielectric relaxation experiments on DMf have been conducted between 238.15 K and 338.15K. The thermophysical properties of the molecular interactions between DMf and the ionic liquids ammonium salts and imidazolium salts have been studied. The characteristics of solute-solvent interactions between ammonium nitrate and DMf have been investigated.