Learn More
Medchemexpress LLC NN1177 (TFA) | 98.7% | 4570.07 (free base) | 25 MG
SDP
Catalog No. 5000426500
Encompass_Preferred
NN1177 TFA is a long-acting GLP-1/glucagon receptor co-agonist. It induces dose-dependent body weight loss in diet-induced obese mice. This product is for research use only.
  • Long-acting GLP-1/glucagon receptor co-agonist
  • Induces dose-dependent body weight loss in diet-induced obese mice
  • Reduces CYP3A4 mRNA expression and activity in human hepatocytes
  • Improves glucose tolerance in diet-induced obese mice
  • Reduces liver fat and inflammatory biomarkers in NASH model mice
Catalog No. 50-004-26500 Supplier Medchemexpress LLC Supplier No. 25MGHYP10032A
NN1177 TFA is a long-acting GLP-1/glucagon receptor co-agonist. It induces dose-dependent body weight loss in diet-induced obese mice. This product is for research use only.
  • Long-acting GLP-1/glucagon receptor co-agonist
  • Induces dose-dependent body weight loss in diet-induced obese mice
  • Reduces CYP3A4 mRNA expression and activity in human hepatocytes
  • Improves glucose tolerance in diet-induced obese mice
  • Reduces liver fat and inflammatory biomarkers in NASH model mice