Learn More
Medchemexpress LLC NN1177 (TFA) | 98.7% | 4570.07 (free base) | 5 MG
SDP
Catalog No. 5000424671
Encompass_Preferred
NN1177 TFA (NNC9204-1177 TFA) is a long-acting GLP-1/glucagon receptor co-agonist. This compound is known to induce dose-dependent body weight loss in diet-induced obese mice and improves glucose tolerance. It has also been shown to reduce liver fat and relevant inflammatory and fibrosis biomarkers in NASH models.
  • Long-acting GLP-1/glucagon receptor co-agonist
  • Induces body weight loss
  • Improves glucose tolerance
  • Reduces liver fat
  • Reduces inflammatory and fibrosis relevant biomarkers
  • For research use only
Catalog No. 50-004-24671 Supplier Medchemexpress LLC Supplier No. HYP10032A5MG
NN1177 TFA (NNC9204-1177 TFA) is a long-acting GLP-1/glucagon receptor co-agonist. This compound is known to induce dose-dependent body weight loss in diet-induced obese mice and improves glucose tolerance. It has also been shown to reduce liver fat and relevant inflammatory and fibrosis biomarkers in NASH models.
  • Long-acting GLP-1/glucagon receptor co-agonist
  • Induces body weight loss
  • Improves glucose tolerance
  • Reduces liver fat
  • Reduces inflammatory and fibrosis relevant biomarkers
  • For research use only