Learn More
Medchemexpress LLC NN1213 acetate | 99.65% | 4553.10 | 100 MG
SDP
Catalog No. 5000424826
Encompass_Preferred
NN1213 acetate is a long-acting human amylin peptide analog that functions as a selective Amylin receptor agonist. This compound significantly reduces food intake and fat mass, and has been shown to decrease body weight in diet-induced obese rats, making it suitable for research related to obesity.
  • Functions as a selective Amylin receptor agonist
  • Exhibits EC50 values of 0.177 nM for hAMY3R and 0.262 nM for rAMY3R
  • Reduces food intake and fat mass
  • Decreases body weight in diet-induced obese rats
  • Suitable for obesity research
Catalog No. 50-004-24826 Supplier Medchemexpress LLC Supplier No. HYP11245A100MG
NN1213 acetate is a long-acting human amylin peptide analog that functions as a selective Amylin receptor agonist. This compound significantly reduces food intake and fat mass, and has been shown to decrease body weight in diet-induced obese rats, making it suitable for research related to obesity.
  • Functions as a selective Amylin receptor agonist
  • Exhibits EC50 values of 0.177 nM for hAMY3R and 0.262 nM for rAMY3R
  • Reduces food intake and fat mass
  • Decreases body weight in diet-induced obese rats
  • Suitable for obesity research