Learn More
Medchemexpress LLC NN1213 acetate | 99.7% | 4553.10 | 5 MG
SDP
Catalog No. 5000422759
Encompass_Preferred
NN1213 acetate is a long-acting human amylin peptide analog and a selective Amylin receptor agonist. It effectively reduces food intake and fat mass, leading to a reduction in body weight in diet-induced obese rats. NN1213 acetate is intended for obesity research.
  • Selective Amylin receptor agonist
  • Reduces food intake and fat mass
  • Reduces body weight in diet-induced obese rats
  • Intended for obesity research
  • Exhibits EC50 values of 0.177 nM for hAMY3R and 0.262 nM for rAMY3R
Catalog No. 50-004-22759 Supplier Medchemexpress LLC Supplier No. HYP11245A5MG
NN1213 acetate is a long-acting human amylin peptide analog and a selective Amylin receptor agonist. It effectively reduces food intake and fat mass, leading to a reduction in body weight in diet-induced obese rats. NN1213 acetate is intended for obesity research.
  • Selective Amylin receptor agonist
  • Reduces food intake and fat mass
  • Reduces body weight in diet-induced obese rats
  • Intended for obesity research
  • Exhibits EC50 values of 0.177 nM for hAMY3R and 0.262 nM for rAMY3R