Learn More
Medchemexpress LLC NN1213 acetate | 99.7% | 4553.10 (free base) | 10 MG
SDP
Catalog No. 5000423428
Encompass_Preferred
NN1213 acetate is a long-acting human amylin peptide analog and a selective amylin receptor agonist. It has EC50 values of 0.177 nM for hAMY3R and 0.262 nM for rAMY3R. This compound significantly reduces food intake and fat mass, leading to body weight reduction in diet-induced obese rats, making it suitable for obesity research.
  • Selective amylin receptor agonist
  • EC50 values of 0.177 nM for hAMY3R and 0.262 nM for rAMY3R
  • Produces dose-dependent body weight loss in diet-induced obese rats
  • Reduces food intake in rats and beagle dogs
  • Decreases body weight and food intake in diet-induced obese rats
Catalog No. 50-004-23428 Supplier Medchemexpress LLC Supplier No. HYP11245A10MG
NN1213 acetate is a long-acting human amylin peptide analog and a selective amylin receptor agonist. It has EC50 values of 0.177 nM for hAMY3R and 0.262 nM for rAMY3R. This compound significantly reduces food intake and fat mass, leading to body weight reduction in diet-induced obese rats, making it suitable for obesity research.
  • Selective amylin receptor agonist
  • EC50 values of 0.177 nM for hAMY3R and 0.262 nM for rAMY3R
  • Produces dose-dependent body weight loss in diet-induced obese rats
  • Reduces food intake in rats and beagle dogs
  • Decreases body weight and food intake in diet-induced obese rats