Learn More
Medchemexpress LLC NN1213 acetate | 99.7% | 4553.10 (free base) | 50 MG
SDP
Catalog No. 5000423995
Encompass_Preferred
NN1213 acetate is a long-acting human amylin peptide analog, functioning as a selective Amylin receptor agonist. It exhibits EC50 values of 0.177 and 0.262 nM for hAMY3R and rAMY3R, respectively. This product significantly reduces food intake and fat mass, and can decrease body weight in diet-induced obese rats, making it suitable for obesity research.
  • Selective Amylin receptor agonist
  • Reduces food intake
  • Reduces fat mass
  • Decreases body weight in diet-induced obese rats
  • Suitable for obesity research
Catalog No. 50-004-23995 Supplier Medchemexpress LLC Supplier No. HYP11245A50MG
NN1213 acetate is a long-acting human amylin peptide analog, functioning as a selective Amylin receptor agonist. It exhibits EC50 values of 0.177 and 0.262 nM for hAMY3R and rAMY3R, respectively. This product significantly reduces food intake and fat mass, and can decrease body weight in diet-induced obese rats, making it suitable for obesity research.
  • Selective Amylin receptor agonist
  • Reduces food intake
  • Reduces fat mass
  • Decreases body weight in diet-induced obese rats
  • Suitable for obesity research