Learn More
Medchemexpress LLC NNC 05-2090 hydrochloride | 184845-18-9 | MFCD00952020 | 99.1% | 451.00 g/mol | C27H31ClN2O2 | 100 MG
SDP
Catalog No. 5000246130
Encompass_Preferred
NNC 05-2090 hydrochloride is a research compound that inhibits GABA uptake and selectively inhibits the mGAT2 (BGT-1) transporter. It exhibits anticonvulsant activity and is used in preclinical studies of epilepsy and other neurological disorders.
  • GABA uptake inhibitor and selective mGAT2 (BGT-1) inhibitor (Ki ≈ 1.4 μM).
  • Demonstrated anticonvulsant activity; used in preclinical epilepsy research.
  • Supplied as the hydrochloride salt with molecular formula C27H31ClN2O2.
  • High purity (99.1% by HPLC) suitable for biochemical and pharmacological studies.
  • Available in multiple pack sizes and as a 10 mM solution in DMSO for in vitro use.
Catalog No. 50-002-46130 Supplier Medchemexpress LLC Supplier No. HY103509100MG
NNC 05-2090 hydrochloride is a research compound that inhibits GABA uptake and selectively inhibits the mGAT2 (BGT-1) transporter. It exhibits anticonvulsant activity and is used in preclinical studies of epilepsy and other neurological disorders.
  • GABA uptake inhibitor and selective mGAT2 (BGT-1) inhibitor (Ki ≈ 1.4 μM).
  • Demonstrated anticonvulsant activity; used in preclinical epilepsy research.
  • Supplied as the hydrochloride salt with molecular formula C27H31ClN2O2.
  • High purity (99.1% by HPLC) suitable for biochemical and pharmacological studies.
  • Available in multiple pack sizes and as a 10 mM solution in DMSO for in vitro use.