Learn More
Medchemexpress LLC NNC-0640 | 307986-98-7 | 98.4% | 573.67 g/mol | C29H31N7O4S | 10MG
SDP
Supplier:  Medchemexpress LLC HY12462210MG
Encompass_Preferred
NNC-0640 is a research-grade negative allosteric modulator of the human glucagon receptor (GCGR), reported to inhibit GCGR with an IC50 of 69.2 nM. It is provided as a high-purity small molecule suitable for in vitro metabolic and diabetes-related studies. For research use only; not for human or clinical use.
  • Negative allosteric modulator of the human glucagon receptor.
  • Reported IC50 of 69.2 nM for GCGR.
  • High purity (≈98.4%).
  • Molecular formula C29H31N7O4S; molecular weight 573.67 g/mol.
  • Supplied in small-molecule research quantities (e.g., 10 MG).
  • Intended for in vitro research use only.
Catalog No. 50-002-36130