Learn More
Medchemexpress LLC HY-124622 5mg Medchemexpress, NNC-0640 CAS:307986-98-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1246225MG
Encompass_Preferred
Medchemexpress, HY-124622 5mg NNC-0640 CAS:307986-98-7 NNC-0640 is a potent human G-protein-coupled glucagon receptor (GCGR) negative allosteric modulator (NAM) with an IC50 of 69.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9672