Learn More
Medchemexpress LLC NNC 38-1049 | 757183-18-9 | 99.8% | 250 MG
SDP
Catalog No. 5000233647
Encompass_Preferred
NNC 38-1049 is a research compound that functions as a potent, orally active, competitive histamine H3 receptor antagonist with reported activity in central histaminergic pathways and effects on feeding behavior.
  • Orally active, potent, competitive histamine H3 receptor antagonist.
  • Increases extracellular histamine levels in the paraventricular nucleus.
  • Shown to decrease food intake and body weight in preclinical studies.
  • Reported purity 99.84%.
  • Chemical formula C19H25ClN2O2; molecular weight 348.87 g·mol⁻¹.
Catalog No. 50-002-33647 Supplier Medchemexpress LLC Supplier No. HY119203250MG
NNC 38-1049 is a research compound that functions as a potent, orally active, competitive histamine H3 receptor antagonist with reported activity in central histaminergic pathways and effects on feeding behavior.
  • Orally active, potent, competitive histamine H3 receptor antagonist.
  • Increases extracellular histamine levels in the paraventricular nucleus.
  • Shown to decrease food intake and body weight in preclinical studies.
  • Reported purity 99.84%.
  • Chemical formula C19H25ClN2O2; molecular weight 348.87 g·mol⁻¹.