Learn More
Medchemexpress LLC NNC 55-0396 | 357400-13-6 | 99.0% | C30H40Cl2FN3O2 | 1 MG
SDP
Catalog No. 502261077
Encompass_Preferred
NNC 55-0396 is a highly selective T-type calcium channel blocker with an IC50 value of 6.8 μM for Cav3.1 T-type channels. It can be utilized for research related to neurological diseases.
  • Highly selective T-type calcium channel blocker
  • IC50 value of 6.8 μM for Cav3.1 T-type channels
  • Useful for neurological disease research
  • Suppresses tremor in GABAA subunit α1-null mice and harmaline mouse models
This item is not returnable. View return policy
Catalog No. 50-226-1077 Supplier Medchemexpress LLC Supplier No. HY507221MG
NNC 55-0396 is a highly selective T-type calcium channel blocker with an IC50 value of 6.8 μM for Cav3.1 T-type channels. It can be utilized for research related to neurological diseases.
  • Highly selective T-type calcium channel blocker
  • IC50 value of 6.8 μM for Cav3.1 T-type channels
  • Useful for neurological disease research
  • Suppresses tremor in GABAA subunit α1-null mice and harmaline mouse models