Learn More
Cayman Chemical NNC 55-0396, Purity: ≥95%, CAS: 357400-13-6, 5 mg
SDP
Catalog No. 502980356
Encompass_Preferred
A selective inhibitor of T-type calcium channels (IC50 = 6.8 μM); exhibits no activity against L-type calcium channels up to 100 μM; inhibits tremor in mice (20 mg/kg and 12.5 mg/kg in GABAA α1-null and harmaline-induced tremor models, respectively); inhibits Kv channels (IC50 = 80 nM)
Catalog No. 50-298-0356 Supplier Cayman Chemical Supplier No. 172165 MG
A selective inhibitor of T-type calcium channels (IC50 = 6.8 μM); exhibits no activity against L-type calcium channels up to 100 μM; inhibits tremor in mice (20 mg/kg and 12.5 mg/kg in GABAA α1-null and harmaline-induced tremor models, respectively); inhibits Kv channels (IC50 = 80 nM)