Learn More
TARGETMOL CHEMICALS INC NNC 63-0532 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T230745MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. NNC 63-0532 is a potentnociceptin receptor agonist Ki=7.3 nM EC50=305 nM. purity: 98%
Catalog No. 50-258-0897