Learn More
Medchemexpress LLC NNC-711 hydrochloride | 145645-62-1 | MFCD00153853 | 99.8% | 386.87 Da | C21H23ClN2O3 | 10 MG
SDP
Catalog No. 5000248638
Encompass_Preferred
NNC-711 (NO-711 hydrochloride) is a potent and selective GAT-1 (GABA transporter 1) inhibitor provided as the hydrochloride salt for research use. It is supplied as a white to off-white solid with high purity and is intended for biochemical and pharmacological studies involving GABA uptake inhibition.
  • Potent and selective GAT-1 inhibitor.
  • Hydrochloride salt, solid form (white to off-white).
  • High purity (99.8%) suitable for research applications.
  • Molecular weight 386.87 Da; formula C21H23ClN2O3.
  • Pack size 10 mg; store sealed at -20°C; in solution store at -80°C for up to 6 months or -20°C for up to 1 month.
Catalog No. 50-002-48638 Supplier Medchemexpress LLC Supplier No. HY10350610MG
NNC-711 (NO-711 hydrochloride) is a potent and selective GAT-1 (GABA transporter 1) inhibitor provided as the hydrochloride salt for research use. It is supplied as a white to off-white solid with high purity and is intended for biochemical and pharmacological studies involving GABA uptake inhibition.
  • Potent and selective GAT-1 inhibitor.
  • Hydrochloride salt, solid form (white to off-white).
  • High purity (99.8%) suitable for research applications.
  • Molecular weight 386.87 Da; formula C21H23ClN2O3.
  • Pack size 10 mg; store sealed at -20°C; in solution store at -80°C for up to 6 months or -20°C for up to 1 month.