Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NNC-711 hydrochloride | 145645-62-1 | MFCD00153853 | 99.8% | 386.87 g/mol | C21H23ClN2O3 | 1 ML
SDP

Catalog No. 5000245331
Encompass_Preferred

NNC-711 (NO-711 hydrochloride) is a potent, selective inhibitor of the GABA transporter GAT-1 used in neuroscience research to modulate GABA uptake. It is supplied as a high-purity solid and as a 10 mM solution for laboratory use.

  • Potent and selective GAT-1 inhibitor.
  • High purity (≈99.8%).
  • Available as solid powder and 10 mM solution.
  • Molecular weight 386.87 g/mol; formula C21H23ClN2O3.
  • Suitable for electrophysiology, GABA uptake assays, and in vivo pharmacology.
  • Provided in research-scale quantities (mg ranges and 1 mL solution).

Catalog No. 50-002-45331 Supplier Medchemexpress LLC Supplier No. HY10350610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NNC-711 (NO-711 hydrochloride) is a potent, selective inhibitor of the GABA transporter GAT-1 used in neuroscience research to modulate GABA uptake. It is supplied as a high-purity solid and as a 10 mM solution for laboratory use.

  • Potent and selective GAT-1 inhibitor.
  • High purity (≈99.8%).
  • Available as solid powder and 10 mM solution.
  • Molecular weight 386.87 g/mol; formula C21H23ClN2O3.
  • Suitable for electrophysiology, GABA uptake assays, and in vivo pharmacology.
  • Provided in research-scale quantities (mg ranges and 1 mL solution).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.