Learn More
Medchemexpress LLC NNC-711 (NO-711 hydrochloride) | 145645-62-1 | MFCD00153853 | 99.8% | 386.87 g/mol | C21H23ClN2O3 | 1 MG
SDP
Catalog No. 5000245332
Encompass_Preferred
NNC-711 (NO-711 hydrochloride) is a potent, selective inhibitor of GAT-1 (GABA transporter 1) used in preclinical research to block GABA uptake. It shows anticonvulsant and analgesic effects in vivo and is supplied as the hydrochloride salt in solid and solution formats for use in in vitro and in vivo pharmacology studies.
  • Potent, selective GAT-1 inhibition (IC50 ≈ 0.04 μM for hGAT-1)
  • High purity (99.8%)
  • Available in small research quantities and solution formats
  • Suitable for in vitro and in vivo pharmacology studies
  • Molecular weight 386.87 g/mol; formula C21H23ClN2O3
Catalog No. 50-002-45332 Supplier Medchemexpress LLC Supplier No. HY1035061MG
NNC-711 (NO-711 hydrochloride) is a potent, selective inhibitor of GAT-1 (GABA transporter 1) used in preclinical research to block GABA uptake. It shows anticonvulsant and analgesic effects in vivo and is supplied as the hydrochloride salt in solid and solution formats for use in in vitro and in vivo pharmacology studies.
  • Potent, selective GAT-1 inhibition (IC50 ≈ 0.04 μM for hGAT-1)
  • High purity (99.8%)
  • Available in small research quantities and solution formats
  • Suitable for in vitro and in vivo pharmacology studies
  • Molecular weight 386.87 g/mol; formula C21H23ClN2O3