Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NNC 92-1687 10mM 1mL | 22903-37-3 | 300.33 g·mol⁻¹ | C15H12N2O3S | 1 ML
SDP

Catalog No. 5000228271
Encompass_Preferred

NNC 92-1687 is a non-peptide competitive antagonist of the human glucagon receptor (GCGR) used as a research tool for studies of glucose homeostasis and type 2 diabetes. It shows IC50 = 20 μM and Ki = 9.1 μM and is supplied as a 10 mM solution in DMSO (1 mL) or as a solid.

  • Non-peptide competitive glucagon receptor antagonist
  • Reported IC50 of 20 μM and Ki of 9.1 μM
  • Molecular formula C15H12N2O3S and molecular weight 300.33 g·mol⁻¹
  • Available as 10 mM solution in DMSO (1 mL) and in multiple solid sizes
  • HPLC purity approximately 98.7%
  • Storage: solid at room temperature (up to 3 years); in solvent at -80°C (2 years) or -20°C (1 year)

Catalog No. 50-002-28271 Supplier Medchemexpress LLC Supplier No. HY11685410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NNC 92-1687 is a non-peptide competitive antagonist of the human glucagon receptor (GCGR) used as a research tool for studies of glucose homeostasis and type 2 diabetes. It shows IC50 = 20 μM and Ki = 9.1 μM and is supplied as a 10 mM solution in DMSO (1 mL) or as a solid.

  • Non-peptide competitive glucagon receptor antagonist
  • Reported IC50 of 20 μM and Ki of 9.1 μM
  • Molecular formula C15H12N2O3S and molecular weight 300.33 g·mol⁻¹
  • Available as 10 mM solution in DMSO (1 mL) and in multiple solid sizes
  • HPLC purity approximately 98.7%
  • Storage: solid at room temperature (up to 3 years); in solvent at -80°C (2 years) or -20°C (1 year)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.