Learn More
Medchemexpress LLC NNC 92-1687 | 1MG
SDP
Catalog No. 5000227612
Encompass_Preferred
NNC 92-1687 is a non-peptide competitive human glucagon receptor antagonist (IC50 20 M Ki 9 1 M) NNC 92-1687 can be used in the research of type 2 diabetes[1]
Catalog No. 50-002-27612 Supplier Medchemexpress LLC Supplier No. HY1168541MG
NNC 92-1687 is a non-peptide competitive human glucagon receptor antagonist (IC50 20 M Ki 9 1 M) NNC 92-1687 can be used in the research of type 2 diabetes[1]