Learn More
TARGETMOL CHEMICALS INC NNC 92-1687 5MG
SDP
Catalog No. 502580538
Encompass_Preferred
Also available in 50 mg 100 mg and bulk. Please contact Fisher for quotes. NNC 92-1687 is a selective glucagon receptor antagonist that can be used to study type 2 diabetes. purity: 99%
Catalog No. 50-258-0538 Supplier TARGETMOL CHEMICALS INC Supplier No. T281825MG
Also available in 50 mg 100 mg and bulk. Please contact Fisher for quotes. NNC 92-1687 is a selective glucagon receptor antagonist that can be used to study type 2 diabetes. purity: 99%