Learn More
Medchemexpress LLC NNC9204-1177 (TFA) | 98.7% | 4570.07 (free base) | 1 MG
SDP
Catalog No. 5000423235
Encompass_Preferred
NN1177 TFA is a long-acting GLP-1/glucagon receptor co-agonist. It can induce a dose-dependent body weight loss in diet-induced obese (DIO) mice and reduces CYP3A4 mRNA expression and activity in human hepatocytes. It can also reduce liver fat and inflammatory and fibrosis biomarkers in a NASH mouse model.
  • Induces dose-dependent body weight loss in diet-induced obese (DIO) mice.
  • Reduces CYP3A4 mRNA expression and activity in human hepatocytes.
  • Reduces liver fat and inflammatory and fibrosis biomarkers in a NASH mouse model.
Catalog No. 50-004-23235 Supplier Medchemexpress LLC Supplier No. HYP10032A1MG
NN1177 TFA is a long-acting GLP-1/glucagon receptor co-agonist. It can induce a dose-dependent body weight loss in diet-induced obese (DIO) mice and reduces CYP3A4 mRNA expression and activity in human hepatocytes. It can also reduce liver fat and inflammatory and fibrosis biomarkers in a NASH mouse model.
  • Induces dose-dependent body weight loss in diet-induced obese (DIO) mice.
  • Reduces CYP3A4 mRNA expression and activity in human hepatocytes.
  • Reduces liver fat and inflammatory and fibrosis biomarkers in a NASH mouse model.