Learn More
Cayman Chemical nngenIn F 2.5mg
SDP
Catalog No. 502991067
Encompass_Preferred
A drimane sesquiterpene; active against B. subtilis (IC50 = 78 UG/ml); cytotoxic to NS-1, DU145, and MCF-7 cancer and NFF non-cancerous cells (IC50s = 49, 95, 49, and 84 UG/ml, respectively)
Catalog No. 50-299-1067 Supplier Cayman Chemical Supplier No. 316582.5 MG
A drimane sesquiterpene; active against B. subtilis (IC50 = 78 UG/ml); cytotoxic to NS-1, DU145, and MCF-7 cancer and NFF non-cancerous cells (IC50s = 49, 95, 49, and 84 UG/ml, respectively)