Learn More
Enzo Life Sciences NNGH (25mg). CAS: 161314-17-6
SDP
Supplier:  Enzo Life Sciences BMLPI1150025
Encompass_Preferred
Potent inhibitor of human stromelysin-1 (MMP-3), Ki=0.13 µM. Also inhibits MMP-1, MMP-2, MMP-7, MMP-8, MMP-9, MMP-10, MMP-12 (metalloelastase), MMP-13, MMP-14, and MMP-20. Alternative name: N-Isobutyl-N-(4-methoxyphenylsulfonyl)glycyl hydroxamic acid. Purity: ≥99% (TLC). Solubility: Soluble in DMSO (30mg/ml) or 100% ethanol (8mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1453