Learn More
Cayman Chemical N-Nitroso-N-methyl-4-Aminobutyric Acid, Purity: ≥95%, CAS: 61445-55-4, 100 mg
SDP
Catalog No. 502991799
Encompass_Preferred
A nitrosamine carcinogen; oxidized to MOPN in isolated rat liver mitochondria; induces bladder transitional cell carcinomas in rats at 300 mg/L per day in the drinking water
Catalog No. 50-299-1799 Supplier Cayman Chemical Supplier No. 30911100 MG
A nitrosamine carcinogen; oxidized to MOPN in isolated rat liver mitochondria; induces bladder transitional cell carcinomas in rats at 300 mg/L per day in the drinking water