Learn More
Cayman Chemical NNItrosoNmethyl4AmInobut 10mg
SDP
Catalog No. 502989882
Encompass_Preferred
A nitrosamine carcinogen; oxidized to MOPN in isolated rat liver mitochondria; induces bladder transitional cell carcinomas in rats at 300 mg/L per day in the drinking water
Catalog No. 50-298-9882 Supplier Cayman Chemical Supplier No. 3091110 MG
A nitrosamine carcinogen; oxidized to MOPN in isolated rat liver mitochondria; induces bladder transitional cell carcinomas in rats at 300 mg/L per day in the drinking water