Learn More
Medchemexpress LLC NNMT-IN-6 hydrochloride | 3031756-18-7 | 98.2% | 544.00 g/mol | C25H30ClN7O5 | 5 MG
SDP
Catalog No. 5000221998
Encompass_Preferred
NNMT-IN-6 hydrochloride is a research-grade small-molecule inhibitor of nicotinamide N-methyltransferase (NNMT). Supplied as the hydrochloride salt for improved solubility and handling, it has reported biochemical potency (IC50 = 1.41 μM; Kd = 5.6 μM). Molecular formula C25H30ClN7O5, molecular weight 544.00, with a COA-reported purity of 98.21%.
  • Potent NNMT inhibitor with reported IC50 = 1.41 μM and Kd = 5.6 μM.
  • Supplied as a hydrochloride salt for improved solubility.
  • High chemical purity appropriate for biochemical and cellular assays.
  • Small-molecule format compatible with standard assay workflows.
  • Available in multiple package sizes to support screening and follow-up studies.
Catalog No. 50-002-21998 Supplier Medchemexpress LLC Supplier No. HY138993A5MG
NNMT-IN-6 hydrochloride is a research-grade small-molecule inhibitor of nicotinamide N-methyltransferase (NNMT). Supplied as the hydrochloride salt for improved solubility and handling, it has reported biochemical potency (IC50 = 1.41 μM; Kd = 5.6 μM). Molecular formula C25H30ClN7O5, molecular weight 544.00, with a COA-reported purity of 98.21%.
  • Potent NNMT inhibitor with reported IC50 = 1.41 μM and Kd = 5.6 μM.
  • Supplied as a hydrochloride salt for improved solubility.
  • High chemical purity appropriate for biochemical and cellular assays.
  • Small-molecule format compatible with standard assay workflows.
  • Available in multiple package sizes to support screening and follow-up studies.