Learn More
Medchemexpress LLC NNMT-IN-6 hydrochloride | 3031756-18-7 | 98.2% | C25H30ClN7O5 | 10MG
SDP
Catalog No. 5000260686
Encompass_Preferred
NNMT-IN-6 hydrochloride is the hydrochloride salt of a small-molecule inhibitor of nicotinamide N-methyltransferase (NNMT) intended for research use. It exhibits in vitro inhibitory activity (IC50 = 1.41 μM; Kd = 5.6 μM) and is supplied as a high-purity solid with documented solubility and storage recommendations for biochemical and cellular assays.
  • High purity (98.2%).
  • Confirmed NNMT inhibitory activity (IC50 = 1.41 μM; Kd = 5.6 μM).
  • Hydrochloride salt form for improved solubility.
  • Validated solubility in DMSO and water (250 mg/mL with sonication).
  • Suitable for biochemical and cellular assays targeting NNMT.
  • Recommended sealed storage away from moisture, with solvent stability guidelines.
Catalog No. 50-002-60686 Supplier Medchemexpress LLC Supplier No. HY138993A10MG
NNMT-IN-6 hydrochloride is the hydrochloride salt of a small-molecule inhibitor of nicotinamide N-methyltransferase (NNMT) intended for research use. It exhibits in vitro inhibitory activity (IC50 = 1.41 μM; Kd = 5.6 μM) and is supplied as a high-purity solid with documented solubility and storage recommendations for biochemical and cellular assays.
  • High purity (98.2%).
  • Confirmed NNMT inhibitory activity (IC50 = 1.41 μM; Kd = 5.6 μM).
  • Hydrochloride salt form for improved solubility.
  • Validated solubility in DMSO and water (250 mg/mL with sonication).
  • Suitable for biochemical and cellular assays targeting NNMT.
  • Recommended sealed storage away from moisture, with solvent stability guidelines.