Learn More
ABclonal Technology NNMT Rabbit PolymAb
SDP
Catalog No. 5000010608
Encompass_Preferred
N-methylation is one method by which drug and other xenobiotic compounds are metabolized by the liver This gene encodes the protein responsible for this enzymatic activity which uses S-adenosyl methionine as the methyl donor
Catalog No. 50-000-10608 Supplier ABclonal Technology Supplier No. A25906PM20UL
N-methylation is one method by which drug and other xenobiotic compounds are metabolized by the liver This gene encodes the protein responsible for this enzymatic activity which uses S-adenosyl methionine as the methyl donor