Learn More
Abcam TMA-DPH, 5MG
Supplier:  Abcam AB2743065MG
Encompass_Preferred
MW 461.6 Da, Purity >95%. Fluorescent probe for membrane fluidity measurements. Acts as an anchor at the lipid/water interface. Specifically and rapidly incorporating into the plasma membrane and between the membranes and the buffer. Especially useful in the study of monolayer dynamics of lipoproteins, exocytosis kinetics and similar systems.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-210