Learn More
Enzo Life Sciences nNOS polyclonal antibody (25 µg)
SDP
Supplier:  Enzo Life Sciences BMLSA2270025
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 1414-1434 (CRSESIAFIEESKKDTDEVFSS) of human brain NOS (nNOS; NOS1). Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat Chicken. Applications: ELISA, ICC, IF, WB. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS containing 0.05% sodium azide.
Catalog No. 50-201-1770