Learn More
Enzo Life Sciences nNOS polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPNO030D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human nNOS. UniProt ID: P29475. Source: Purified from rabbit serum. Species reactivity: Mouse, Rat. Applications: IHC (PS), WB. Recommended Dilutions: Western Blot (1:250, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7284