Learn More
Roboz Surgical NO.1 DUMONT DUMOSTAR
SDP
Catalog No. NC9091197
Encompass
Dumont Tweezers; Pattern #1; .20 X .12mm Tip Size; 120mm Length; Material: Dumostar
Catalog No. NC9091197 Supplier Roboz Surgical Supplier No. RS4960
Dumont Tweezers; Pattern #1; .20 X .12mm Tip Size; 120mm Length; Material: Dumostar