Learn More
Ace Glass, Inc. #13 PTFE Stopper with Polypropylene Extraction Nut, Solid
SDP
Catalog No. 50977614
Encompass
#13 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.
Catalog No. 50-977-614 Supplier Ace Glass, Inc. Supplier No. 1263632
#13 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.