Learn More
Chemglass Life Sciences PTFE Seal only, fits GL-14 Cap
SDP
Catalog No. CG19610
Encompass_Preferred
PTFE Seal only, fits GL-14 Cap. Cap is made from PBT (thermoplastic polyester) and will withstand temperatures from -45 to 200º C. PTFE seal is also available separately.
Catalog No. CG19610 Supplier Chemglass Life Sciences Supplier No. CG19610
PTFE Seal only, fits GL-14 Cap. Cap is made from PBT (thermoplastic polyester) and will withstand temperatures from -45 to 200º C. PTFE seal is also available separately.