Learn More
Ace Glass, Inc. Complete 32mm OD vacuum trap, #15 threaded side arm and innertube
SDP
Catalog No. 50976318
Encompass_Preferred
32mm O.D. vacuum trap consists of body with Ace-Thred, adjustable inner tube, and nylon bushing with FETFE o-ring. Has #15 threaded side arm. For use with PTFE Ace-Safe connectors, to allow safe, easy tubing connect-disconnect.
Catalog No. 50-976-318 Supplier Ace Glass, Inc. Supplier No. 874615
32mm O.D. vacuum trap consists of body with Ace-Thred, adjustable inner tube, and nylon bushing with FETFE o-ring. Has #15 threaded side arm. For use with PTFE Ace-Safe connectors, to allow safe, easy tubing connect-disconnect.