Learn More
Ace Glass, Inc. Side arm adapter, 105°, #15 Ace-Thred top, 45/50 inner joint bottom, 45/50 outer joint side
SDP
Catalog No. 50971685
Encompass_Preferred
105 degree side arm adapter with #15 Ace-Thred at top, 45/50 standard taper inner joint at bottom, and 45/50 standard taper outer joint on the side.
Catalog No. 50-971-685 Supplier Ace Glass, Inc. Supplier No. 505096
105 degree side arm adapter with #15 Ace-Thred at top, 45/50 standard taper inner joint at bottom, and 45/50 standard taper outer joint on the side.